missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 420535000

 View more versions of this product

Catalog No. AC420535000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hydroxide pentahydrate
62.0°C to 71.0°C
Authentic
Glass bottle
(CH3)4NOH·5H2O
500 g
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium;hydroxide;pentahydrate
82620
98%
10424-65-4
White
98%
C4H23NO6
MFCD00149566
04, 50
Solubility in water: soluble.
O.O.O.O.O.[OH-].C[N+](C)(C)C
181.23
181.23
Crystalline Powder

Chemical Identifiers

10424-65-4
181.23
MYXKPFMQWULLOH-UHFFFAOYSA-M
82620
O.O.O.O.O.[OH-].C[N+](C)(C)C
C4H23NO6
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
tetramethylazanium;hydroxide;pentahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life w

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only