missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethylammonium Hydroxide Pentahydrate 97.0+%, TCI America™
$97.76 - $1320.68
Chemical Identifiers
CAS | 10424-65-4 |
---|---|
Molecular Formula | C4H23NO6 |
Molecular Weight (g/mol) | 181.23 |
MDL Number | MFCD00149566 |
InChI Key | MYXKPFMQWULLOH-UHFFFAOYSA-M |
Synonym | tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm |
PubChem CID | 82620 |
IUPAC Name | tetramethylazanium pentahydrate hydroxide |
SMILES | O.O.O.O.O.[OH-].C[N+](C)(C)C |
Chemical Identifiers
10424-65-4 | |
181.23 | |
MYXKPFMQWULLOH-UHFFFAOYSA-M | |
82620 | |
O.O.O.O.O.[OH-].C[N+](C)(C)C |
C4H23NO6 | |
MFCD00149566 | |
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm | |
tetramethylazanium pentahydrate hydroxide |
Specifications
10424-65-4 | |
White | |
MFCD00149566 | |
3423 | |
MYXKPFMQWULLOH-UHFFFAOYSA-M | |
tetramethylazanium pentahydrate hydroxide | |
82620 | |
≥97.0% (T) | |
Tetramethylammonium Hydroxide Pentahydrate |
73°C | |
C4H23NO6 | |
25 g | |
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm | |
O.O.O.O.O.[OH-].C[N+](C)(C)C | |
181.23 | |
91.15 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186
RTECSNumber : PA0875000
TSCA : Yes