missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylcyclobutane-1,3-dione, 99%

CAS: 933-52-8 | C8H12O2 | 140.182 g/mol

$90.56 - $1045.05

Chemical Identifiers

CAS 933-52-8
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001331
InChI Key RGCDVHNITQEYPO-UHFFFAOYSA-N
Synonym tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
PubChem CID 13617
IUPAC Name 2,2,4,4-tetramethylcyclobutane-1,3-dione
SMILES CC1(C(=O)C(C1=O)(C)C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1629709
View Documents
Thermo Scientific Chemicals
A1629709
10 g
Each for $90.56
Only null left
 
AAA1629718
View Documents
Thermo Scientific Chemicals
A1629718
50 g
Each for $282.45
Only null left
 
AAA1629730
View Documents
Thermo Scientific Chemicals
A1629730
250 g
Each for $1,045.05
Only null left
 
Description

Description

Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.

Solubility
It is soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
Specifications

Chemical Identifiers

933-52-8
140.182
RGCDVHNITQEYPO-UHFFFAOYSA-N
13617
CC1(C(=O)C(C1=O)(C)C)C
C8H12O2
MFCD00001331
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
2,2,4,4-tetramethylcyclobutane-1,3-dione

Specifications

933-52-8
120°C to 122°C (150 mmHg)
MFCD00001331
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4
RGCDVHNITQEYPO-UHFFFAOYSA-N
2,2,4,4-tetramethylcyclobutane-1,3-dione
13617
99%
112°C to 115°C
C8H12O2
10 g
It is soluble in chloroform.
CC1(C(=O)C(C1=O)(C)C)C
140.182
140.18
Tetramethylcyclobutane-1,3-dione
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 213-269-6

RTECSNumber : GU1775000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only