missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylphosphonium bromide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1790706

 View more versions of this product

Catalog No. AAA1790706


Explore available promotions

Description

Description

It is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

Tetramethylphosphonium bromide
C4H12BrP
5 g
tetramethylphosphonium bromide, tetramethylphosphanium bromide, acmc-1ai8x, ch3 4pbr, tetramethyl phosphonium bromide, phosphonium, tetramethyl-, bromide
ZTXFOCMYRCGSMU-UHFFFAOYSA-M
tetramethylphosphanium;bromide
357594
98%
4519-28-2
MFCD00011802
Hygroscopic
Insoluble in water.
[Br-].C[P+](C)(C)C
171.02
171.03

Chemical Identifiers

4519-28-2
171.02
ZTXFOCMYRCGSMU-UHFFFAOYSA-M
357594
[Br-].C[P+](C)(C)C
C4H12BrP
MFCD00011802
tetramethylphosphonium bromide, tetramethylphosphanium bromide, acmc-1ai8x, ch3 4pbr, tetramethyl phosphonium bromide, phosphonium, tetramethyl-, bromide
tetramethylphosphanium;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only