missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylsilane, ACS reagent, NMR grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 445900250

 View more versions of this product

Catalog No. AC445900250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetramethylsilane
-99°C
0.6400g/mL
-27°C
C4H12Si
Si(CH3)4
25 g
15,9377
Solubility in water: 0.02g/L (25°C). Other solubilities: soluble in most common organic solvents
C[Si](C)(C)C
88.23
CHEBI:85361
ACS Reagent
75-76-3
Colorless
26°C to 28°C
Glass bottle
1.3585
MFCD00008274
0.64
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
CZDYPVPMEAXLPK-UHFFFAOYSA-N
tetramethylsilane
6396
88.22
Liquid

Chemical Identifiers

75-76-3
88.23
CZDYPVPMEAXLPK-UHFFFAOYSA-N
6396
tetramethylsilane
C4H12Si
MFCD00008274
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073
CHEBI:85361
C[Si](C)(C)C
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 200-899-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only