missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyltin, 98%, Thermo Scientific Chemicals

$146.01 - $658.90

Chemical Identifiers

CAS 594-27-4
Molecular Formula C4H12Sn
Molecular Weight (g/mol) 178.83
MDL Number MFCD00008278
InChI Key VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Synonym tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
PubChem CID 11661
ChEBI CHEBI:30420
IUPAC Name tetramethylstannane
SMILES C[Sn](C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Qty  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC163980100
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
163980100
10 g Glass bottle
Each for $146.01
 
AC163980500
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
163980500
50 g Glass bottle
Each for $658.90
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

594-27-4
178.83
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
11661
tetramethylstannane
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
CHEBI:30420
C[Sn](C)(C)C

Specifications

594-27-4
Colorless
74.0°C to 75.0°C
Authentic
Glass bottle
1.4400 to 1.4420
MFCD00008278
04, 631
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
tetramethylstannane
11661
178.83
Liquid
-54.0°C
1.2900g/mL
−12°C
97.5% min. (GC)
C4H12Sn
(CH3)4Sn
10 g
1.29
Solubility in water: immiscible.
C[Sn](C)(C)C
178.83
CHEBI:30420
98%
Tetramethyltin, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal if inhaled.
Fatal in contact with skin.
Highly flammable liquid and vapor.
Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Rinse mouth.

GHS Signal Word: Danger

EINECSNumber : 209-833-6

RTECSNumber : WH8630000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only