Learn More
Tetraphenylboron sodium, ACS reagent, Thermo Scientific Chemicals
$235.10 - $3,137.70
Chemical Identifiers
CAS | 143-66-8 |
---|---|
Molecular Formula | C24H20BNa |
Molecular Weight (g/mol) | 342.22 |
MDL Number | MFCD00011494 |
InChI Key | HFSRCEJMTLMDLI-UHFFFAOYSA-N |
Synonym | sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride |
PubChem CID | 2723787 |
IUPAC Name | sodium tetraphenylboranuide |
SMILES | [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC424450250
|
Thermo Scientific Chemicals
424450250 |
25 g | Glass bottle |
N/A
|
|
||||
AC424451000
|
Thermo Scientific Chemicals
424451000 |
100 g | Glass bottle |
N/A
|
|
||||
AC424455000
|
Thermo Scientific Chemicals
424455000 |
500 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
143-66-8 | |
342.22 | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
2723787 | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20BNa | |
MFCD00011494 | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
sodium tetraphenylboranuide |
Specifications
>300.0°C | |
Colorless to White | |
25 g | |
(C6H5)4BNa | |
15,8818 | |
Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
342.22 | |
342.21 | |
Authentic | |
Glass bottle |
143-66-8 | |
Crystals or Granular Powder | |
C24H20BNa | |
MFCD00011494 | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
sodium tetraphenylboranuide | |
2723787 | |
ACS Reagent | |
0.5 % max. (105°C) | |
Tetraphenylboron sodium |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Danger
EINECSNumber : 205-605-5
RUO – Research Use Only