missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetraphenylcyclopentadienone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138540050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetraphenylcyclopentadienone | |
479-33-4 | |
Black | |
98.5% min. (HPLC) | |
C29H20O | |
5 g | |
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one | |
68068 | |
99% |
99% | |
217.0°C to 222.0°C | |
Authentic | |
Glass bottle | |
MFCD00001407 | |
01,1149; 04,490; 06,577 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
384.47 | |
384.47 | |
Fine Crystalline Powder |
Chemical Identifiers
479-33-4 | |
384.47 | |
PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
68068 | |
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
C29H20O | |
MFCD00001407 | |
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
Safety and Handling
LIGHT SENSITIVE
EINECSNumber : 207-530-3
TSCA : TSCA
RUO – Research Use Only