missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraphenylcyclopentadienone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 138540250

 View more versions of this product

Catalog No. AC138540250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetraphenylcyclopentadienone
479-33-4
Black
98.5% min. (HPLC)
C29H20O
25 g
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone
PLGPSDNOLCVGSS-UHFFFAOYSA-N
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one
68068
99%
99%
217.0°C to 222.0°C
Authentic
Glass bottle
MFCD00001407
01,1149; 04,490; 06,577
Solubility in water: insoluble.
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5
384.47
384.47
Fine Crystalline Powder

Chemical Identifiers

479-33-4
384.47
PLGPSDNOLCVGSS-UHFFFAOYSA-N
68068
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5
C29H20O
MFCD00001407
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 207-530-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only