missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrapropyl Orthosilicate 98.0+%, TCI America™
$48.46 - $328.08
Chemical Identifiers
| CAS | 682-01-9 |
|---|---|
| Molecular Formula | C12H28O4Si |
| Molecular Weight (g/mol) | 264.437 |
| MDL Number | MFCD00009364 |
| InChI Key | ZQZCOBSUOFHDEE-UHFFFAOYSA-N |
| Synonym | tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane |
| PubChem CID | 69627 |
| IUPAC Name | tetrapropyl silicate |
| SMILES | CCCO[Si](OCCC)(OCCC)OCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T117425ML
|
TCI America
T117425ML |
25 mL |
Each for $48.46
|
|
|||||
|
T1174500ML
|
TCI America
T1174500ML |
500 mL |
Each for $328.08
|
|
|||||
Chemical Identifiers
| 682-01-9 | |
| 264.437 | |
| ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
| 69627 | |
| CCCO[Si](OCCC)(OCCC)OCCC |
| C12H28O4Si | |
| MFCD00009364 | |
| tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
| tetrapropyl silicate |
Specifications
| 682-01-9 | |
| 225°C | |
| MFCD00009364 | |
| 1760 | |
| ZQZCOBSUOFHDEE-UHFFFAOYSA-N | |
| tetrapropyl silicate | |
| 69627 | |
| ≥98.0% (GC) | |
| Tetrapropyl Orthosilicate |
| Colorless | |
| C12H28O4Si | |
| 25 mL | |
| tetrapropoxysilane, tetrapropyl orthosilicate, tetra-n-propoxysilane, propyl orthosilicate, silicic acid h4sio4 , tetrapropyl ester, unii-4pe821g3gh, tetra-n-propyl orthosilicate, propyl silicate, dynasil p, tetrapropoxy silane | |
| CCCO[Si](OCCC)(OCCC)OCCC | |
| 264.437 | |
| 264.44 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2048
TSCA : Yes