missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrapropylammonium bromide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 209375000

 View more versions of this product

Catalog No. AC209375000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrapropylammonium bromide
1941-30-6
White
97.5 to 102.5% (Argentometry)
C12H28BrN
MFCD00011840
04,I,364
Solubility in water: 100g/L water
CCC[N+](CCC)(CCC)CCC.[Br-]
266.26
CHEBI:55318
98%
98%
266°C to 272°C
Authentic
Plastic bottle
(CH3CH2CH2)4NBr
500 g
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
BGQMOFGZRJUORO-UHFFFAOYSA-M
tetrapropylazanium;bromide
74745
266.26
Crystalline Powder or Crystals

Chemical Identifiers

1941-30-6
266.26
BGQMOFGZRJUORO-UHFFFAOYSA-M
74745
tetrapropylazanium;bromide
C12H28BrN
MFCD00011840
tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
CHEBI:55318
CCC[N+](CCC)(CCC)CCC.[Br-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-727-6

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only