missing translation for 'onlineSavingsMsg'
Learn More

Tetrasodium Ethylenediaminetetraacetate Tetrahydrate 98.0+%, TCI America™

$44.73 - $98.79

Chemical Identifiers

CAS 13235-36-4
Molecular Formula C10H20N2Na4O12
Molecular Weight (g/mol) 452.23
MDL Number MFCD00150027
InChI Key XFLNVMPCPRLYBE-UHFFFAOYSA-J
Synonym tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
PubChem CID 23287279
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
SMILES O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E056525G
View Documents
TCI America
E056525G
25 g
Each for $44.73
Only null left
 
E0565500G
View Documents
TCI America
E0565500G
500 g
Each for $98.79
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13235-36-4
452.23
XFLNVMPCPRLYBE-UHFFFAOYSA-J
23287279
O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
C10H20N2Na4O12
MFCD00150027
tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate

Specifications

13235-36-4
C10H20N2Na4O12
25 g
XFLNVMPCPRLYBE-UHFFFAOYSA-J
tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate tetrahydrate
23287279
≥98.0% (T)
Tetrasodium Ethylenediaminetetraacetate Tetrahydrate
White
MFCD00150027
tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
452.23
380.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents