Decon™ The Pill™ Autoclave Deodorizer
Neutralizes odors from autoclaving
Supplier:  Decon™ DC5100
 View more versions of this product
Catalog No. 04-355-23
Explore available promotions
Description
Description
  • One capsule treats 7 cu. ft. of autoclave capacity
  • Gelatin capsule keeps deodorizer off fingers, ensures economical dose rate
Specifications
Specifications
Clear Amber
Autoclavable
100 Capsules