Decon™ The Pill™ Autoclave Deodorizer
Neutralizes odors from autoclaving
Supplier: Decon™ 5500
Description
- One capsule treats 7 cu. ft. of autoclave capacity
- Gelatin capsule keeps deodorizer off fingers, ensures economical dose rate
Specifications
Clear Amber | |
Autoclavable | |
500 Capsules |