missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Theobromine, 99%, Thermo Scientific Chemicals

Theobromine, CAS # 83-67-0, is a methylxanthine alkaloid that is present in large quantities in cocoa and is a derivative of caffein.

Supplier:  Thermo Scientific Chemicals 258822500

 View more versions of this product

Catalog No. AC258822500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Theobromine is a methylxanthine alkaloid that is a derivative of caffeine

Applications
• Theobromine is a phosphodiesterase inhibitor, weak adenosine receptor antagonist, and smooth muscle relaxant
• As a catabolic product of caffeine, it has antioxidant and prooxidant properties
• It can be used as a standard for quantification of theobromine

Specifications

Specifications

Theobromine
83-67-0
White
Authentic
Plastic bottle
MFCD00022830
26, 2336
theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
Solubility in water: slightly soluble. Other solubilities: moderately soluble in ammonia, practically insoluble in benzene, ether, carbon, tetrachloride and chloroform
CN1C=NC2=C1C(=O)NC(=O)N2C
180.17
CHEBI:28946
99%
99%
290.0°C to 295.0°C
0.5% max. (1 g, 105°C)
99%
C7H8N4O2
250 g
15, 9433
290.0 °C
YAPQBXQYLJRXSA-UHFFFAOYSA-N
3,7-dimethylpurine-2,6-dione
5429
180.17
Fine Powder

Chemical Identifiers

83-67-0
180.17
YAPQBXQYLJRXSA-UHFFFAOYSA-N
5429
3,7-dimethylpurine-2,6-dione
C7H8N4O2
MFCD00022830
theobromine, 3,7-dimethylxanthine, diurobromine, teobromin, theosalvose, theostene, santheose, thesodate, thesal, theobromin
CHEBI:28946
CN1C=NC2=C1C(=O)NC(=O)N2C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 201-494-2

RTECSNumber : XH2275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only