Therapak™ Aqui-Pak™ Absorbent Sleeves
Ideal for Biological Substance and Exempt Human specimen shipments
$473.39 - $516.70
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
22-130-018
|
Therapak™
10314 |
Up to four tubes |
Pack of 300 Each for $473.39
Case of 3 Pack for $1,420.17
|
|
|||||
22-130-043
|
Therapak™
10316 |
Up to six tubes |
Pack of 300 Each for $516.70
Case of 3 Pack for $1,550.10
|
|
|||||
Description
Ideal for Biological Substance and Exempt Human specimen shipments
- For safe transport of specimen tubes in accordance with DOT/IATA regulations
- Consist of multilayered polymeric material with extremely high absorption properties
- Simultaneously separates and cushions specimen tubes
- Place between primary and secondary container
- Roll it up and place it in a bag or canister
Specifications
Absorbent Sleeve |