Therapak™ Foil-Top Collection Kits
Available in single- or split-specimen collection kits
$794.97 - $1184.88
Specifications
For Use With (Application) | For drugs of abuse testing |
---|
Includes: All kits include one 8 oz. collection cup with pour spout and temperature strip; one 45mL tamper-evident, flip-top transport vial graduated at 15 and 30mL; and a transport bag with absorbent material. Split-specimen kit, designed for testing that requires two samples from a single donor, includes two transport vials.
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Product Type | Price | Quantity | |||||
22-130-410
|
Therapak™
39449 |
8 oz. Collection cup with pour spout, temperature strip, two 45 mL tamper-evident, flip-top transport vials, transport bag with absorbent | Split Foil-top Collection Kit |
Case of 100 Each for $1,184.88
|
|
|||||
22-130-409
|
Therapak™
39448 |
8 oz. Collection cup with pour spout, temperature strip, one 45 mL tamper-evident, flip-top transport vials, transport bag with absorbent | Single Foil-top Collection Kit |
Case of 100 Each for $794.97
|
|
|||||
Description
- For drugs of abuse testing applications
- Kit contents conveniently sealed inside the collection cup with peel-off foil lid
Specifications
For drugs of abuse testing |