Therapak™ Insulated Envelope and Gel Wrap
Provides excellent thermal insulation for transport of temperature-sensitive items
$102.36 - $632.76
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | ||||||
22-130-121
|
Therapak™
TR56422 |
Gel Wrap |
Case of 16 Each for $102.36
|
|
||||||
22-130-122
|
Therapak™
56424 |
Insulated Envelope |
Case of 32 Each for $455.85
|
|
||||||
22-130-413
|
Therapak™
37981 |
Envelope/Gel Wrap Combo w/Box |
Case of 32 Each for $632.76
|
|
||||||
Description
As a shipping system, the insulated envelope/gel wrap combo (wrap is housed inside the envelope as a secondary barrier) prevents short-duration (2-3 hours) temperature spikes encountered during transport. Envelope and gel wrap may be purchased separately, or as a system.
Insulated Envelope
- Reusable bag with resealable zip-seal closure and leak-resistant inner compartment
- Restricts penetration of thermal energy; reflects 99% of all radiant heat transfer
- Minimizes shock and vibration of specimens during transit
- Ideal choice for transporting ambient specimens in a cool to room temperature environment
- 0°C (32°F) gel formulation insulates specimen tubes
Specifications
For temperature-sensitive items |