missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Therapak™ Metal Duramark™ Lock Boxes

Supplier:  Therapak™ 36746

 View more versions of this product

Catalog No. 22-131-453


Explore available promotions

Description

Description

  • 24 Gauge cold rolled steel construction
  • Painted with powder coat finish for excellent strength and durability
  • Insulated on inside with foam assisting preserving the integrity of the specimens
  • With hinged front door that rotates down 90° for convenient access and use as temporary tray
  • Dimensions: 14 × 10 1/2 × 7 in.
  • Supplied with lock and two matching keys for security
  • Standard color: White
  • Standard printing: Reflex blue text, red biohazard symbol on front side only
  • Optional straps: Under the door, over the door
  • Customization: Any deviation from the standard stock configuration, which may include changes in color, size, design features, closure and print

Specifications

Specifications

Standard Metal Lock Box with No Strap
24 gauge cold rolled steel construction
Videos
SDS
Documents

Documents

Promotions

Promotions