Learn More
Therapak™ Prostate Biopsy - Histology Cassette Collection and Transport Kit
To collect and transport biopsy tissue to diagnose prostate cancer, neoplasm, the cause of prostate enlargement and other prostate related abnormalities
Supplier: Therapak™ 38872
Description
Tissue biopsies are commonly used in pathology laboratory testing to diagnose disease. Once the biopsy specimen collected in accordance with established protocols, each preprinted biopsy specimen cassette can be placed into the color coded prefilled formalin vial. Once the completed test requisition is placed into the provided biohazard bag with absorbent, the kit is ready for transport to the laboratory via a preferred courier. Packing and shipping instructions are located in each kit.
- Feature preprinted biopsy location identification
- Reduces error and saves time
- Two large prefilled color-coded formalin vials marked for Left (L) and Right (R) tissue transport
Compliance
Complies with IATA regulations for safe transport to the laboratory for testing
Specifications
Complies with IATA regulations for safe transport | |
Collect and transport biopsy tissue to diagnose prostate cancer, neoplasm, the cause of prostate enlargement and other prostate related abnormalities | |
20/Cs. |
Prostate Biopsy - Histology Cassette Collection and Transport Kit | |
Preprinted histology cassettes; Color coded prefilled formalin vials; Biohazard bag with absorbent |