Therapak™ Refrigerated Biological Substance Category B Shipping Systems
Certified for ground and air shipments of Biological Substance, Category B specimens.
Supplier: Therapak™ 37814
Includes: All systems include transport box with UN3373 Biological Substance, biohazard and orientation arrow labels; EPS foam cooler, gel refrigerant pack(s), 95kPa bags or canisters, a variety of inner packaging choices (depending on the type and and quantity of specimens being transported), absorbent material, list of contents card, cushioning material, and packing instructions.
Description
- DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens
- Systems suitable for overnight shipments; Mfr. No. 38028 field-tested for 2-8°C temperature maintenance for 48 hours of transport
Specifications
Holds 18 Tubes | |
Transport box, medium cooler, large 95kPa canister, three Aqui-Pak™ segmented absorbent pouches, 8 oz. refrigerant packs, cushioning material, list of contents card and required labels for IATA PI650 marking and compliance. | |
DOT and IATA Packing Instruction 650 compliant for the shipment of refrigerated specimens |