Therapak™ StoneScreen™ Collection and Transport Kit
Collect and transport kidney stone samples for laboratory analysis
Supplier: Therapak™ 38879
Description
The kit features kidney stone collection cup constructed to capture renal calculi and for use with patients attempting to pass stones following extracorporeal shock wave lithotripsy. Once the stone is captured, it can be placed in the storage space under the lid and saved for analysis. After the entire cup and sample is re-collapsed following collection, the specimen cup is placed into the zip style bag and is returned into the transport box along with the test requisition for courier pick up.
- Features collapsible stone collection cup and lid with sample storage compartment
- Plastic strainer cup includes nylon screen at the bottom
- Collection instructions included with each kit
Specifications
Kidney Stone Collection and Transport Kit | |
Capture renal calculi from patients attempting to pass stones following extracorporeal shock wave lithotripsy | |
16/Cs. |
StoneScreen Collection and Transport Kit | |
Collapsible collection cup and lid with sample storage compartment; Zip-style bag; Transport box |