Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers
Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
Supplier:  Thermo Scientific™ 2124*
 View more versions of this product
Catalog No. 11-670-4C
Explore available promotions
Description
Description
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
Stainless Steel:
  • Durable, corrosion-resistant stainless-steel exterior
  • Carrying handle and vented secure lid (Part numbers 2122, 2123, 2124)
Shallow Wide Mouth:
  • Ideal for use as a cold trap
  • Convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol
  • Stainless-steel jacket and cushioned base for years of trouble-free use
  • Without lid and handle (Part number 2129)
Specifications
Specifications
With carrying handle and vented secure lid
4.5 L
Benchtop
182 mm
4.5 kg
Benchtop LN2 Container
Stainless Steel
345 mm
150 mm
Videos
SDS
Documents
Documents
Accessories