Learn More
Thermo Scientific™ Tubes, 0.2 mL, flat cap, purple
0.2 mL individual tubes with attached flat or dome snap shut caps, available in a variety of colors
Supplier: Thermo Scientific™ AB0620P
Description
0.2 mL Individual Tubes with integral “snap shut” caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
The Thermo Scientific 0.2 mL Individual Tubes with integral ”snap shut“ caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when closed
•
Integral 'snap shut' cap
• Available with flat or domed caps
Applications
• PCR
• Sample storage
Hazardous:No
>Storage Condition:Ambient
PCR; Sample storage
Certifications
Certified free from human DNA, RNases, and DNases
Specifications
Certified DNA/RNase Free | |
0.2 mL | |
1000 tubes | |
Polypropylene | |
Flat Caps | |
Tube |
Purple | |
0.25 mL | |
Thermal Cyclers | |
Standard | |
1,000 Tubes |