Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
Supplier:  Thermo Scientific™ B100038I
 View more versions of this product
Catalog No. B100038I
Explore available promotions
Description
Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
  • Minimizes bioburden due to construction under sterile cGMP conditions
  • Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
130 L
Store at room temperature.
This product is packaged and sealed into plastic dust cover packaging which is then placed into carboard cartons labeled with product and lot identification.
SV30112
ASI 26
2D Tank Liner System
45.7 cm x 76.2 cm (18 in. x 30 in.)
20 per case
Irradiated
Tubing