missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Abgene™ Plate Covers

Polypropylene 96-well plate covers, with or without ridges for easy stacking

Supplier:  Thermo Scientific™ AB0755

Catalog No. AB-0755


Explore available promotions

Description

Description

Polypropylene 96-well plate covers, with or without ridges for easy stacking

Thermo Scientific Polypropylene Plate Covers with stacking ridges provide effective sealing options for a variety of applications and can be used with 96-well plates.

Highlights

  • Ridged covers for stacking of covered plates
  • Non-ridged covers for use with automated systems
  • Polypropylene covers, ridged and non-ridged, are available for use with and 384-well plates

Hazardous: No

Storage Condition: Ambient

Specifications

Specifications

Natural
5 x 3.31 x 0.36 in. (126 x 84 x 9.25mm)
Cleanroom
With stacking ridge
50
5 x 3.31 in. (126 x 84 mm)
Microplate Lid, cover
None
All well shapes
H1 Corner
Microplate Lid
Polypropylene
96
No
2
Lid
Abgene
-20°C to +120°C
Abgene 96 and 384-well Plate
PCR Ready
Videos
SDS
Documents

Documents

Promotions

Promotions