Learn More
Thermo Scientific™ Abgene Storage Plate Caps, Strips of 8
Storage Plate Caps and Mats are available in a variety of formats.
Supplier: Thermo Scientific™ AB0981
Description
Storage Plate Caps and Mats are available in a variety of formats.
Thermo Scientific storage plate cap strips are suitable for use on with Round Well Storage Plates and feature a low-profile design. They come in an 8-cap strip and can simultaneously seal up to 8 well. Each cap is individually numbered.
Highlights
- 8-cap strip
- Each cap is individually numbered
- Simultaneous sealing of 8 wells
- End tabs for ease of removal
- Low profile for increased well volume
Autoclavable- Resistance: Not Recommended
DMSO (100%) - Resistance: Successfully Tested
Ethanol (100%) - Resistance: Successfully Tested
Gamma Irradiation - Resistance: Successfully Tested
Hazardous: No
Isopropanol (100%) - Resistance: Successfully Tested
Long Term Strorage: Successfully Tested
Peelable: Successfully Tested
Pierceable:Not Recommended
Resealable: Successfully Tested
Sealing Temp Range: -20°C to +55°C
Storage Condition: Ambient
UV Irradiation- Resistance: Successfully Tested
Specifications
Natural | |
Cleanroom | |
8 | |
Abgene | |
No | |
Abgene 96 Round Well Plate |
Polypropylene | |
Autoclavable | |
Plate Lid | |
Yes | |
Round |