Thermo Scientific™ ART™ Non-Filtered Pipette Tips
Range of styles and packaging options
Supplier: Thermo Scientific™ MB3901
Description
Range of styles and packaging options
- All sterile HLT branded tips are certified “Pure”
- All nonsterile tips are manufactured in clean room environments and are certified DNA, RNase and DNase free
- Stringent manufacturing and QA/QC testing procedures assure the most aseptic products on the market
- Some tips have reference marks which provide a quick and convenient guide to visually establish whether you are pipeting the desired volume of liquid
Specifications
200 μL | |
Sterile | |
DNase, RNase, DNA, ATP and Endotoxin Free | |
Standard | |
Sterile |
Non-filtered | |
Yellow | |
Rack | |
ART |