missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Citadel™ Tissue Processor Accessories

For use with the Epredia™ Citadel Tissue Processor

Supplier:  Epredia™ 69810053

Catalog No. 69810053


Explore available promotions

Description

Description

Specifications

Specifications

Wax Bath Assembly
Tissue Processor
Ea.
Videos
SDS
Documents

Documents

Promotions

Promotions