Epredia™ Disposable Blade Carriers
Utilize a quick clamp system that accepts low- and high-profile blades
Supplier:  Epredia™ 705830
Catalog No. 36-102-2568
Explore available promotions
Description
Description
Updated models (long stroke and version 2 bar guideways) include removable angle adjustment and blade clamp levers.
Updated Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
  • Only compatible with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
Standard Disposable Blade Carriers
  • Quick clamp system for high- and low-profile blades
  • Clearance angle is adjustable
  • Upper part moves laterally without opening the blade clamp
  • Moveable knife guard covers the blade entirely in any position
  • Complete with knife carrier base
Specifications
Specifications
Long Stroke Units