missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Gene Frame, 25 μL (1.0 x 1.0 cm)

Gene Frames for in-situ feature a dual strength adhesive system which facilitates the easy removal of the frames after use.

Supplier:  Thermo Scientific™ AB0576

Catalog No. AB-0576

Explore available promotions

Includes: 100 Rigid coverslips for easy handling and uniform hybridizations

Description

Description

Thermo Scientific Gene Frames have adhesives on either side of the frame which are of differing strengths; the side which adheres to the glass has an easy release adhesive whereas the reverse, which sticks to the coverslip, forms a stronger bond. This means that when the coverslip is removed, due to the disparity in the strengths of the different adhesives, the frame is removed simultaneously with no sticky residue being left behind on the slide.
Highlights
• Gas tight sealing prevents reagent loss due to evaporation
• Can withstand temperatures of 97°C
• Designed for use with standard microscope slides
• Frame and coverslip can be removed simultaneously leaving no residue
• Can be autoclaved or sterilized by UV irradiation
• All Gene Frame packs are supplied with 100 coverslips
• Rigid coverslips for ease of handling and uniform hybridizations
• Thickness of Gene Frames: 0.25 mm
Applications
• In-situ PCR

Hazardous:No
>Storage Condition:Ambient

Certifications

Certified free from DNase, RNase and human genomic DNA.

Specifications

Specifications

100 Coverslips
0.25 mL
Microarray Slide Frame
0.25 mm
Universal
Gene Frame
Videos
SDS
Documents

Documents

Promotions

Promotions