Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Cylindrical Longpath Cell Holder
Supplier:  Thermo Scientific™ 336012000
Catalog No. 14-385-400
Explore available promotions
Description
Description
For GENESYS 10 Spectrophotometers with single-cell cuvet holder.
  • Accommodates a cylindrical cell with a 10 to 100mm pathlength, 22 to 25mm dia. and Z dimension of 11mm Models with six-position cell holders can accommodate single-cell holders using accessory Single-cell Platform and Holder (14-385-476), available separately
Specifications
Specifications
Cylindrical Cell Holder
Genesys™ 10 UV-vis spectrophotometer
100 mm