Thermo Scientific™ GENESYS™ 10 UV-Vis Spectrophotometers Test Tube Holder
Supplier:  Thermo Scientific™ 336014000
Catalog No. 14-385-401
Explore available promotions
Description
Description
  • Accommodates one 25 dia. × 102mmH test tube or COD reagent vial
  • For GENESYS 10 spectrophotometers with six-position cell holder (holds three) or single-cell holder platform (holds one)
Specifications
Specifications
Test Tube Holder
Genesys™ 10 UV-vis spectrophotometer
One 25 dia. x 102 mm H test tube or COD reagent vial