Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
$271.25 - $655.54
Specifications
For Use With (Equipment) H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor
View More Specs
Products 8
Catalog Number Mfr. No. Capacity Price Quantity  
Catalog Number Mfr. No. Capacity Price Quantity  
00-385
View Documents
Thermo Scientific™
00385
108 x 15 mL Blood Collection Tube
Each for $527.54
00-436
View Documents
Thermo Scientific™
00436
30 x 50 mL Tubes
Each for $655.54
00-444
View Documents
Thermo Scientific™
00444
6 x 500 mL Bottles
Each for $314.29
00-445
View Documents
Thermo Scientific™
00445
18 x 50 mL Tubes
Each for $561.85
00-511
View Documents
Thermo Scientific™
00511
450 mL Blood Bag
Each for $273.13
00-447
View Documents
Thermo Scientific™
00447
48 x 15 mL Conical Blood Collection Tube
Each for $561.85
00-512
View Documents
Thermo Scientific™
00512
6 x 550 mL Blood Bags
Each for $271.25
00-857
View Documents
Thermo Scientific™
00857
5 mL Blood Collection Tubes
Each for $533.15
Description
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
Specifications
Specifications
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'