Thermo Scientific™ MBP Polystyrene Test Tubes
Catalog No. 02-681-201
Explore available promotions
Description
Description
Porex Polystyrene Test Tubes
Note: Caps are not included; sold separately.
Specifications
Specifications
75 mm
Single-use
1000
Test Tubes
Polystyrene
Test tubes. (Caps are sold separately.)
12 mm