missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters

Use these calibration standards to check your colorimeter instrument operations before measuring COD chemistries, for example in environmental and wastewater monitoring.

Supplier:  Thermo Scientific™ ORICODS10

 View more versions of this product

Catalog No. 13-642-046


Explore available promotions

Description

Description

Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.

  • For use with: Orion AQUAfast AQ4000, AQ3140 and AQ2040 colorimeters
  • Two Calibration Standards are available: 1000 ppm (CODS01) and 10000 ppm (CODS10), both in 475 mL bottles
  • Ideal for environmental and wastewater monitoring

Order Info

Reagents sold separately.

Specifications

Specifications

10000ppm
AQUAfast Colorimeter
Environmental and Wastewater Monitoring
475 mL
EPA
Videos
SDS
Documents

Documents

Promotions

Promotions