Thermo Scientific™ Polypropylene Centrifuge Bottles
Polypropylene bottle with cap
Supplier:  Thermo Scientific™ 75007583
 View more versions of this product
Catalog No. 75-007-583
Explore available promotions
Description
Description
Polypropylene bottle with cap
Specifications
Specifications
13 oz., 400mL
13 oz.
Centrifuges
400 mL
Polypropylene