missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ RevertAid RT Reverse Transcription Kit

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates.

Supplier:  Thermo Scientific™ K1691

Catalog No. FERK1691


Explore available promotions

Description

Description

The Thermo Scientific RevertAid RT Kit is a complete system for efficient synthesis of first strand cDNA from mRNA or total RNA templates. The kit uses RevertAid Reverse Transcriptase (RT), which has lower RNase H activity compared to AMV reverse transcriptase. The enzyme maintains activity at 42–50°C and is suitable for synthesis of cDNA up to 13 kb. The recombinant RiboLock RNase Inhibitor, supplied with the kit, effectively protects RNA from degradation at temperatures up to 55°C.

First strand cDNA synthesized with this system can be directly used as a template in PCR or real-time PCR. It is also ideal as a first step for second strand cDNA synthesis or linear RNA amplification. Radioactively and non-radioactively labeled nucleotides can be incorporated into first strand cDNA for use as a probe in hybridization experiments, including microarrays.

Features of the RevertAid RT Kit include:
• Full-length first strand cDNA up to 13 kb
• Optimum reaction temperature 42°C
• Complete kit includes all the components for RT reactions

Applications
• First strand cDNA synthesis for RT-PCR and RT-qPCR
• Construction of full length cDNA libraries
• Antisense RNA synthesis

Order Info

Orders placed Monday through Wednesday ship next day; Thursday and Friday orders ship the following Monday.

Specifications

Specifications

• RevertAid Reverse Transcriptase
• Ribolock RNase Inhibitor
• 5X Reaction Buffer
• dNTP Mix
• Random Hexamer Primer
• Nuclease-free water

Store at –20°C.


60 min.
42°C
RevertAid
Dry Ice
First-Strand cDNA
Separate components
Up to 13 kb
Kit
Reverse Transcription
500 reactions
Yes
Real Time PCR (qPCR)
500 Reactions
Reverse Transcription
RNA
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.