missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Richard-Allan Scientific™ Masson Trichrome Kit

Aids in differentiating collagen and smooth muscle in tumors. Comes with all stains and solutions necessary to complete the staining procedure.

Supplier:  Epredia™ 87019

Catalog No. 22-110-648


Explore available promotions

Includes: Kit stains red for muscle, cytoplasm and keratin, blue for collagen and mucin and black for nuclei

Description

Description

Masson Trichrom stain kit stains red for muscle, cytoplasm and keratin, blue for collagen and mucin and black for nuclei 


Kit Components:

  • Aniline Blue Stain Solution - 250mL (8.4 oz.)
  • Biebrich Scarlet Acid Fuchsin Solution - 250mL (8.4 oz.)
  • Bouin’s Fluid - 500mL (16.9 oz.)
  • Phosphotungstic-Phosphomolybdic Acid Solution (250mL (8.4 oz.)
  • Weigerts Iron Hematoxylin, Part A - 500mL (16.9 oz.)
Specifications

Specifications

Masson Trichrome Kit
For detection of connective and muscle tissue pathologie
Kit
Videos
SDS
Documents

Documents

Promotions

Promotions