missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ Shandon™ Pathcentre™ Tissue Processor Accessories
For use with the Epredia™ Shandon™ Tissue Processor
Supplier: Epredia™ 66110048
Specifications
Airbag | |
Tissue Processor | |
4 Pk. |