Learn More
Epredia™ Shandon™ Rapid-Chrome H & E Frozen Section Staining Kit
Produces permanently stained slides
Supplier: Epredia™ 9990001
Includes: Rapid-Fixx fixative, deionized water, Eosin Y, Xylene, Gill 3 Hematoxylin, bluing reagent, 100% alcohol, mounting medium
Description
- Contains Rapid-Fixx™ alcohol-base formalin ideal for fixing slides from frozen sections
Specifications
| Stain Kit | |
| Ideal for fixing slides from frozen sections | |
| Rapid-Fixx fixative, deionized water, Eosin-Y, xylene, Gill 3 Hematoxylin, bluing reagent, 100% alcohol and mounting medium, as well as five slide staining grips | |
| 1 Kit |