Thermo Scientific™ SUN-SRi™ Dissolution Cannulae
Angled for easy access to samples
$276.92 - $278.81
Catalog Number | Mfr. No. | For Use With | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Price | Quantity | ||||||
14-823-277
|
Thermo Scientific™
401350 |
900 mL, 4 3/8 in. Needle |
N/A
|
|
||||||
14-823-278
|
Thermo Scientific™
401352 |
500 mL, 7 1/2 in. Needle |
N/A
|
|
||||||
Description
- Type 316 stainless steel
- Luer-LOK™ connection for easy attachment
- Tip sleeve allows secure filter attachment
Order Info
All plastic syringes sold separately
Specifications
Cannula |