Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
$2502.36 - $3711.38
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
11-670-5A
|
Thermo Scientific™
TY509X1 |
5 L |
Each for $2,502.36
|
|
|||||
11-670-5B
|
Thermo Scientific™
TY509X2 |
10 L |
Each for $2,614.84
|
|
|||||
11-670-5C
|
Thermo Scientific™
TY509X3 |
20 L |
Each for $3,134.98
|
|
|||||
11-670-5D
|
Thermo Scientific™
TY509X4 |
32 L |
Each for $3,711.38
|
|
|||||
Description
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
- Designed to bring liquid nitrogen to other cryovessels
- Lightweight aluminum dewars assure low-static evaporation rates
- Optional withdrawal device, 12mL dipper and wheeled accessory cart
- Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
- Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
- Warranty varies by country, please contact us for details
Specifications
LN2 Transfer Vessel |