Sonoco™ ThermoSafe Horizontal Bottle Shippers
Horizontal rectangular-shaped EPS foam containers
$382.00 - $488.61
Specifications
Type Bottle Shipper
Material Expanded Polystyrene Foam
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity  
03-530-68
View Documents
Sonoco™
361UPS
473 mL 21.8 x 11 x 11 cm
Pack of 64 Each for $382.00
03-530-69
View Documents
Sonoco™
393UPS
900 mL 27 x 14 x 14 cm
Case of 45 Each for $488.61
Description
Description
  • No outer mailing cartons required—pressure-sensitive mailing labels adhere directly to foam
  • Apply tape to molded grooves for easy sealing
  • Accepted by UPS, parcel post, and other common carriers
Compliance
UN/DOT approved
Specifications
Specifications
Bottle Shipper
Expanded Polystyrene Foam
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'