Sonoco™ ThermoSafe Insulated EPS with Corrugated Containers and Coolers (Medium)
Large selection of cost-effective, reusable EPS foam in corrugated stock containers available in small, medium, and large sizes. EPS foam with corrugated shippers for the safe storage and delivery of most refrigerated or frozen products.
Supplier:  Sonoco™ 662
 View more versions of this product
Catalog No. 03-001-109
Explore available promotions
Description
Description
  • Sizes that range 11 inches-17 inches in length
  • Also available in ThermoSafe Insulated EPS with Corrugated Containers and Coolers- Small (6  inches-10.5 inches in length)
  • Also available in ThermoSafe Insulated EPS with Corrugated Containers and Coolers- Large (19 inches-24 inches in length)
  • 200 lb. test corrugated
  • Excellent dry ice sublimation… average dry ice usage in 1-1/2 inch thick wall units is 5 pounds over 24 hours and 3-4 pounds thereafter.
  • Standard 1-1/2 inch wall shippers are ideal for refrigerated shipments for 24-48 hours.
Specifications
Specifications
4 per carton
14 in.
Safe storage and delivery of refrigerated or frozen material (pharmaceuticals, laboratory specimens, serums, vaccines, human organs, culture pack and nuclear and isotope materials)
12 in.
14.25 X 14.25 X 11.5 in.
12 in.
3 lb.
<24 hr.
12 X 12 X 9.75 in.
15.25 in.
Rounded Corners and Edges
11.5 in.
9.75 in.
24.76 cm
1.13 in.
91kg Test Corrugated Carton
1.4 kg
Molded Expanded Polystyrene