Sonoco™ ThermoSafe Laminated Fiberboard Storage/Transport Units
Lightweight, low-cost mobile storage units. For transport of a wide variety of laboratory, field collection, and field applications. Ideal for dry ice or wet ice storage.
$109.70 - $109.70
Products 1
Catalog Number Mfr. No. Capacity (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Price Quantity  
03-530
View Documents
Sonoco™
314
24 lb.
Each for $109.70
Pack of 8 Each for $877.60
Description
Description
  • Lightwieght laminated fiberboard wtih EPS foam interior.
  • Low-temperature storage chests include 124.7kg (275 lb.) bursting test laminated fiberboard exterior and expanded polystyrene foam interior; with insulated lid.
  • Units are low maintenance and reusable.
  • Available in different sizes for different holding time.
  • Straps, harness, and dollies are sold separately.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'