missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sonoco™ ThermoSafe Medical Transporter Totes
Ideal for onsite collection, storage, and transportation of temperature-sensitive specimens, medications, blood products, medical devices, and organs
$756.85 - $811.91
Specifications
Wall Thickness (English) | 1/4 in. |
---|---|
Description | Includes removable, rigid thermoformed tray-ideal for washing and reusing. Prevents leaking inside and out of transport unit; reinforces durability |
Material | Washable, weather resistant nylon shell with polyethylene foam insulation |
Catalog Number | Mfr. No. | Capacity (English) | Size | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Size | Price | Quantity | ||||
03-531-105
|
Sonoco™
932 |
0.53 cu. ft. | Wall thickness: 0.38 in. |
Each for $811.91
|
|
||||
03-531-104
|
Sonoco™
TS931 |
0.3 cu. ft. | Wall thickness: 0.25 in. |
Each for $756.85
|
|
||||
Description
- Nylon exterior
- Removable, rigid thermoformed tray facilitates cleaning and prevents leaks outside of the tote
- Tray reinforces tote durability
- Adjustable nylon shoulder carrying strap
Specifications
1/4 in. | |
Includes removable, rigid thermoformed tray-ideal for washing and reusing. Prevents leaking inside and out of transport unit; reinforces durability | |
Washable, weather resistant nylon shell with polyethylene foam insulation |