Sonoco™ ThermoSafe Nomadic™ Pre-Qualified Shippers: Accessories
For protection of refrigerated and frozen pharmaceuticals and biological samples and specimens
$8679.88 - $8679.88
Specifications
Applications | For refrigerated, frozen pharmaceutical and biological specimens |
---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
22-031-871
|
Sonoco™
E280C |
Shipper; 2-8 °C; 48-hour |
Case of 12 Each for $8,679.88
|
|
|||||
Description
- Meet toughest ambient profiles of any premium shipper in the industry
- Multicomponent packing system to ensure exceptional temperature protection and durability
- Model E48UPS:
- Complies with international clean air regulations, CFC- and HCFC-free
- Qualified for maintaining payloads of a single 1mL vial to 3L of liquid
- Reusable
- Replaceable components available separately
- With two double-wall laminated pads
- Model E24AUPS:
- Specifically engineered for quick trips of 24 hours or less
- Lightweight and cost-effective EPS inner shell reduces overnight shipping costs
- Superior insulation power of 2 in. thick walls
- With one double-wall laminated pad
Order Info
Container, corepacks and gels shipped in a single overpack.
Specifications
For refrigerated, frozen pharmaceutical and biological specimens |