Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
$111.70 - $411.12
Catalog Number | Mfr. No. | Dimensions (LxWxH) | Weight (English) | Weight (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (LxWxH) | Weight (English) | Weight (Metric) | Price | Quantity | |||
03-530-011
|
Sonoco™
FPP15 |
4.5 x 4 x 1.5 in. (11.4 x 45.72 x 3.81 cm) | 15 oz. | 0.43 kg |
Pack of 36 Each for $126.03
|
|
|||
03-531-53
|
Sonoco™
FPP18 |
8.25 x 10 x 0.75 in. (20.9 x 20.3 x 1.9 cm) | 18 oz. | 0.72 kg |
Pack of 28 Each for $154.61
|
|
|||
03-531-55
|
Sonoco™
FPP24 |
6.5 x 4.5 x 1.5 in. (16.51 x 11.43 x 3.81 cm) | 24 oz. | 0.68 kg |
Pack of 24 Each for $131.42
|
|
|||
03-530-012
|
Sonoco™
FPP31 |
9 x 4 x 1.5 in. (22.8 x 10.16 x 3.81 cm) | 31 oz. | 1.2 kg |
Case of 18 Each for $111.70
|
|
|||
03-600-250
|
Sonoco™
FPP1000 |
4.5 x 4 x 0.5 in. (11.4 x 10.1 x 1.3 cm) | 5 oz. | 142 g |
Pack of 2 Each for $411.12
|
|
|||
03-530-013
|
Sonoco™
FPP64 |
10.5 x 6.5 x 1.75 in. (26.7 x 15.24 x 4.4 cm) | 64 oz. | 1.81 kg |
Case of 10 Each for $122.91
|
|
|||
03-530-010
|
Sonoco™
FPP7B |
4.5 x 4 x 0.75 in. (11.4 x 53.34 x 1.9 cm) | 7.5 oz. | 0.21 kg |
Pack of 72 Each for $223.73
|
|
|||
Description
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
- Available in a broad selection of styles and sizes
- Maintain temperature for an extended time
- Never change shape while in transit
- Freeze at 0°C (32°F)
- Nontoxic