Sonoco™ ThermoSafe U-tek™ Gel Pack
Phase Change Material (PCM) gel pack for shipping temperature-sensitive materials.
Supplier:  Sonoco™ 427
 View more versions of this product
Catalog No. 03-525-7G
Explore available promotions
Description
Description
  • Gel will phase change at -1 deg. or -23 deg.
  • Maintains critical temperatures of serums, test samples, live or frozen tissues, organs, and pharmaceuticals for extended periods.
  • Laminated, metallized film exterior provides radiant heat barrier and resists punctures.
  • Thermal solutions for temperature critical packages during transit
  • condition coolant in freezer below -1 deg. or -23 deg. before shipments.
  • For frozen or refrigerated shipments.
  • Alternative to dry ice.
  • Can be paired with our Mini Mailer(440).
  • Available in several sizes
Specifications
Specifications
6/Cs.
1.18 in.
-23°C
10.5 in.
Laminated, Metallized Film
1361 g
19.5 cm
U-tek Refrigerant Pack
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
2.9 cm
26.7 cm
48 oz.
7.75 in.
10.5 x 7.75 x 1.18 in.